Dextran, M.W. ~ 40,000, Powder

Supplier: SPECTRUM CHEMICAL MFG CORP

DE124-100GM DE124-25KGBL DE124-25GM DE124-1KG
700013-114EA 457.01 USD
700013-114 700013-116 700013-118 700013-120
Dextran, M.W. ~ 40,000, Powder
Dextran

Dextran, 40,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR