Caprinaldehyde ≥95% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Decylaldehyde, Decanal, Decyl aldehyde

D2771-25GM D2771-100GM
700003-694EA 75.95 USD
700003-694 700003-692
Caprinaldehyde ≥95% for synthesis
Caprinaldehyde

Decanal, also known as Decyl aldehyde or caprinaldehyde, is an organic compound and the simplest ten-carbon aldehyde which is used in flavoring and fragrances.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 207
Melting Pt: 7
Density: 0.830
Storage Temperature: Ambient
MDL Number: MFCD00007031
CAS Number: 112-31-2
EINECS: 203-957-4

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR