1,4-Dimethoxybenzene ≥98%
Supplier: Spectrum Chemical Mfg. Corp.
p-Dimethoxybenzene, also known as Hydroquinone dimethyl ether or p-Methoxyanisole, is an organic compound used in soaps and perfumes; it is also an intermediate in synthesis of certain pharmaceuticals.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₈H₁₀O₂ Boiling Pt: 212.6 °C (1013 hPa) Melting Pt: 54…56 °C Density: 0.874 g/cm³ (24.9 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008401 CAS Number: 150-78-7 EINECS: 205-771-9 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...