Dodecyltrimethylammonium chloride solution_SC 50% alcohol, high_purity

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

D1072-200LTB D1072-20LTBL D1072-100ML D1072-4LT
700007-590EA 7268.05 USD
700007-590 700007-592 700007-588 700007-594
Dodecyltrimethylammonium chloride solution_SC 50% alcohol, high_purity
Dodecyltrimethylammonium chloride

Dodecyltrimethylammonium Chloride, 50 Percent Solution in Alcohol, High Purity is a quaternary ammonium compound (QAC) commonly used as a cationic surfactant.


The High Purity grade denotes that this chemical is the highest quality commercially available. Spectrum Chemical manufactured High Purity grade products meet the toughest regulatory standards for quality and purity.

Formula: C₁₅H₃₄ClN
MW: 263.89 g/mol
Melting Pt: 248…249.5 °C
Density: 0.88
Storage Temperature: Ambient
MDL Number: MFCD00041974
CAS Number: 112-00-5
EINECS: 203-927-0
UN: 1219
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR