1-Dodecanol ≥98% ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol

D1067-20LT D1067-3KG D1067-100GM D1067-200LTB D1067-500GM
700002-722EA 1459.85 USD
700002-722 700002-724 700002-718 700002-720 700002-726
1-Dodecanol ≥98% ACS for synthesis
1-Dodecanol

Dodecyl Alcohol, Reagent, ACS is an organic compound that is a colorless, solid and has a floral scent. It is used to produce lubricating oils, pharmaceuticals and surfactants. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: H₃C(CH₂)₁₀CH₂OH
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 22…24 °C
Density: 0.833
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004753
CAS Number: 112-53-8
EINECS: 203-982-0
UN: 3077
ADR: 9,III
Merck Index: 13,03439

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR