Cetrimonium bromide ≥98% for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
CE122-50KGBL
CE122-125GM
CE122-500GM
CE122-2.5KG
700001-278EA
8154
USD
700001-278
700001-272
700001-276
700001-274
Cetrimonium bromide ≥98% for synthesis
Cetrimonium bromide
Cetyltrimethylammonium Bromide is also referred to as CTAB and is a cationic surfactant that is used as a buffer solution during the extraction of DNA, in hair conditioning products, as well as, in the synthesis of gold nanoparticles.
Formula:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Melting Pt: 237…243 °C Flash Pt: 244 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00011772 CAS Number: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III Merck Index: 13,02034 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...