1-Hexadecanol ≥90% NF suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Cetyl alcohol, Palmityl alcohol
CE120-2.5KG
CE120-500GM
CE120-12KG
95032-478EA
486.42
USD
95032-478
95032-480
89078-706
1-Hexadecanol ≥90% NF suitable for use as excipient
1-Hexadecanol
Cetyl Alcohol, NF is used as an emulsion stabilizer, opacifier, emollient and surfactant.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
H₃C(CH₂)₁₄CH₂OH Boiling Pt: 305…330 °C (1013 hPa) Melting Pt: 46…52 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004760 CAS Number: 36653-82-4 EINECS: 253-149-0 Merck Index: 13,02037 |
Specification Test Results
Assay | 90% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...