Oil of cedar wood

Supplier: Spectrum Chemical Mfg. Corp.
Warning

CE108-100ML CE108-500MLG
700010-578EA 97.7 USD
700010-578 700010-580
Oil of cedar wood
Oil of cedar wood
Density: 0,952
Storage Temperature: Ambient
MDL Number: MFCD00146327
CAS Number: 8000-27-9

Order Now

Specification Test Results

Refractive index @20°C 1.510-1.520


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR