Chromium(III) chloride hexahydrate ≥98% USP

Supplier: Spectrum Chemical Mfg. Corp.
Warning

C1344-500GM C1344-12KGBL C1344-50KGBL C1344-125GM
95032-568EA 1312.4 USD
95032-568 89078-730 89078-732 95032-566
Chromium(III) chloride hexahydrate ≥98% USP
Chromium (III) chloride hexahydrate

Chromic Chloride, Hexahydrate, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CrCl₃·6H₂O
Melting Pt: 85 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 13,02242

Order Now

Specification Test Results

Assay 98% Min.
INSOLUBLE MATTER max. 0.01%
IRON (Fe) max. 0.01%
SULFATES (SO4) max. 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR