Citric acid monohydrate 99,5, powder_crystalline USP suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
C1296-500GM
C1296-12KG
C1296-2.5KG
C1296-50KGBL
95032-608EA
255.29
USD
95032-608
89078-746
95032-606
89078-748
Citric acid monohydrate 99,5, powder_crystalline USP suitable for use as excipient
Citric acid monohydrate
Citric Acid, Monohydrate, Crystalline Powder, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₈O₇.H₂O MW: 210.14 g/mol Melting Pt: 135…152 °C Density: 1.552 g/cm³ (20 °C) Flash Pt: 173.9 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 EINECS: 201-069-1 Merck Index: 13,02350 |
|
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...