Calcium fluoride ≥97%, powder, Reagent Grade
Supplier: Spectrum Chemical Mfg. Corp.
|
Warning
|
Calcium Fluoride, Powder, Reagent, an inorganic compound, occurs as the mineral fluorite or fluorspar. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
CaF₂ Boiling Pt: 2500 °C (1013 hPa) Melting Pt: 1418 °C Density: 3.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 13,01669 |
Specification Test Results
Assay | 97% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...