Orthoboric acid ≥99.5%, powder NF
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Boric Acid, Powder, NF is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
B(OH)₃ Boiling Pt: 185 °C (1013 hPa) Melting Pt: 160 °C Density: 1.44 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 13,01326 |
|
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...