Bismuth(III) nitrate basic ≥79%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate

BI150-2.5KG BI150-500GM BI150-125GM
95030-866EA 2109.7 USD
95030-866 95030-868 95030-864
Bismuth(III) nitrate basic ≥79%, powder USP
Bismuth (III) nitrate basic

Bismuth Subnitrate, Powder, USP is used as a dermatological agent and antidiarrheal. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: Bi₅O(OH)₉(NO₃)₄
MW: 1461.99 g/mol
Density: 4.928 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064844
CAS Number: 1304-85-4
EINECS: 215-136-8
UN: 1477
ADR: 5.1 II

Order Now

Specification Test Results

Assay 79% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR