Bentonite, powder NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Montmorillonite, Bentonite

BE125-12KG BE125-2.5KG BE125-500GM BE125-22.5KG BE125-22.5KG
89078-596EA 1160 USD
89078-596 95031-966 95031-968 700000-342 89078-598
Bentonite, powder NF suitable for use as excipient
Bentonite

Bentonite, Powder, NF is used as suspending and emulsifying agent. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Density: 2.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130611
CAS Number: 1302-78-9
EINECS: 215-108-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR