Benzyl butyrate ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Butanoic acid benzyl esterButyric acid benzyl ester

B2805-100ML B2805-500ML
700004-812EA 245.84 USD
700004-812 700004-810
Benzyl butyrate ≥98%
Benzyl butyrate

Benzyl Butyrate is a perfuming agent with a fruity, sweet, tropical, plum, pear, pineapple, apricot scent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 240 °C (1013 hPa)
Density: 1.009
Storage Temperature: Ambient
MDL Number: MFCD00027133
CAS Number: 103-37-7
EINECS: 203-105-1

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR