Orthoboric acid ≥99.5%, powder ACS
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Boric Acid, Powder, Reagent, ACS is a weak acid of boron often used as an antiseptic, insecticide, flame retardant, neutron absorber, or precursor to other chemical compounds. It exists in the form of colorless crystals or a white powder that dissolves in water. Boric acid is poisonous if taken internally or inhaled in large quantities. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness
Formula:
B(OH)₃ Boiling Pt: 185 °C (1013 hPa) Melting Pt: 160 °C Density: 1.44 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 13,01326 |
|
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...