Benzalkonium chloride solution 50% NF
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Benzalkonium Chloride Solution, 50 Percent, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilizing agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇) Melting Pt: 29…34 °C Density: 0.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00145757 CAS Number: 63449-41-2 EINECS: 264-151-6 UN: 2920 ADR: 8.3,II |
Specification Test Results
Assay (%) | 47.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...