Antimony(III) potassium oxitartrate trihydrate ≥99%, powder USP
Supplier: Spectrum Chemical Mfg. Corp.
|
Warning
|
Antimony Potassium Tartrate, Powder, USP is also referred to as emetic tartar. It has long been known as a powerful emetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₈H₄K₂O₁₂Sb₂·3H₂O Melting Pt: 100 °C Density: 2.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167056 CAS Number: 28300-74-5 EINECS: 229-436-1 UN: 1551 ADR: 6.1,III Merck Index: 13,00706 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...