p-Anisaldehyde 98, reagent for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Methoxybenzaldehyde

AN122-500MLG AN122-100ML AN122-5ML
700003-820EA 259.48 USD
700003-820 700003-818 700003-822
p-Anisaldehyde 98, reagent for synthesis
p-Anisaldehyde

p-Anisaldehyde, Reagent, also known as 4-Anisaldehyde, is considered a useful stain in thin layer chromatography. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 140
Melting Pt: 0…2 °C
Density: 1.119
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR