4-Aminobenzoic acid ≥98.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
PABA
, Vitamin Bx
, para-Aminobenzoic acid
, Vitamin H1
AM150-50KGBL
AM150-2.5KG
AM150-100GM
95031-780EA
982.26
USD
95031-780
95031-778
700007-226
4-Aminobenzoic acid ≥98.5% USP
4-Aminobenzoic acid
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 340 °C (1013 hPa) Melting Pt: 188 °C Density: 1.45 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007894 CAS Number: 150-13-0 EINECS: 205-753-0 Merck Index: 13,00422 |
Specification Test Results
Assay | 98.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...