Acetic acid glacial ≥99.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Methanecarboxylic acid
, Vinegar acid
, Ethanoic acid
AC110-2.5LTG
AC110-4CS4
AC110-4LTPV
AC110-200LTB
AC110-2.5PC4
AC110-20LTBL
AC110-2.5LTG
AC110-500MLG
AC110-100ML
AC110-20LTPL
AC110-500CS6
AC110-500MLG
AC110-2.5GC4
AC110-2.5LTP
AC110-2.5LTP
AC110-200LTB
700004-292EA
204
USD
700004-292
95031-384
95031-386
700004-290
700004-284
95031-380
95031-382
95031-426
95031-422
95031-388
95031-424
89078-558
89078-556
700004-288
95031-390
700004-286
Acetic acid glacial ≥99.5% USP
Acetic acid
Glacial Acetic Acid, USP does have some antibacterial and antifungal properties but is used primarily as a pH adjuster. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 118 °C (1013 hPa) Melting Pt: 17 °C Density: 1.049 Flash Pt: 38.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8.3,II REACH: 01-2119475328-30 |
Specification Test Results
Assay | 99.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...