4-Methoxybenzyl alcohol ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Anise alcohol
, 4-(Hydroxymethyl)anisole
, (4-Methoxyphenyl)methanol
, Anisyl alcohol
, p-Anisyl Alcohol
p-Anisyl Alcohol is used as a pharmaceutical intermediate and has a slightly sweet aroma of anise and commonly used in the preparation of jasmine, lilac and other flavors and perfume ingredients.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: 20…24 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004653 CAS Number: 105-13-5 EINECS: 203-273-6 Merck Index: 13,00608 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...