Tetrachlorosilane ≥99.9999% (Si basis) (50ppm-Fe), PURATREM fiber optic grade

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Silicon tetrachloride, Silicon (IV) chloride

98-0147-100G 98-0147-500G
100203-650EA 152.86 USD
100203-650 100211-310
Tetrachlorosilane ≥99.9999% (Si basis) (50ppm-Fe), PURATREM fiber optic grade
Tetrachlorosilane

Halides for CVD & ALD

Formula: Cl₄Si
Boiling Pt: 57.6 °C
Melting Pt: –70 °C
Density: 1.483
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 12,08644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR