Lead(II) chloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lead dichloride
93-8206-500G
93-8206-100G
100201-152EA
212.31
USD
100201-152
100207-906
Lead(II) chloride ≥99%
Lead (II) chloride
Formula:
Cl₂Pb MW: 278.11 g/mol Boiling Pt: 950 °C Melting Pt: 501 °C Density: 5.85 Storage Temperature: Ambient |
MDL Number:
MFCD00011157 CAS Number: 7758-95-4 EINECS: 231-845-5 UN: 2291 ADR: 6.1,III Merck Index: 13,05422 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...