Thallium(III) trifluoroacetate, Technical Grade
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Thallic trifluoroacetate
93-8137-25G
93-8137-5G
100202-582EA
704.86
USD
100202-582
100202-584
Thallium(III) trifluoroacetate, Technical Grade
Thallium (III) trifluoroacetate
Formula:
C₆F₉O₆Tl Boiling Pt: 160 °C (1.5 mmHg) Melting Pt: 169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000414 CAS Number: 23586-53-0 EINECS: 245-761-1 UN: 1707 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...