Platinum ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
Synonyms:
Platinium
93-7832-1G
93-7832-5G
100207-500EA
366.58
USD
100207-500
100203-588
Platinum ≥99.9%, powder
Platinum
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Density: 21.45 Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 UN: 3089 ADR: 4 III Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...