Tungsten ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-7455-50G 93-7455-250G
100199-186EA 46.71 USD
100199-186 100207-160
Tungsten ≥99.9%, powder
Tungsten
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19.3
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4 III
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR