Tungsten ≥99.95%, wire

Supplier: STREM CHEMICALS INC MS
Warning

93-7450-25MT 93-7450-5MT
100210-904EA 619.94 USD
100210-904 100210-556
Tungsten ≥99.95%, wire
Tungsten
Formula: W
Boiling Pt: 5660 °C
Melting Pt: 3410 °C
Density: 19.3
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR