Tungsten ≥99.95%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-7437-50G
93-7437-250G
100207-902EA
43.88
USD
100207-902
100205-762
Tungsten ≥99.95%, powder
Tungsten
Formula:
W Boiling Pt: 5660 °C Melting Pt: 3410 °C Density: 19,3 |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...