Hafnium ≥99.6% (Zr-2.2%), powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-7227-10G
93-7227-2G
93-7227-50G
100197-268EA
154.28
USD
100197-268
100205-750
100207-148
Hafnium ≥99.6% (Zr-2.2%), powder
Hafnium
Formula:
Hf MW: 178.49 Boiling Pt: 4602 °C Melting Pt: 2227 °C Density: 13.31 |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 UN: 2545 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...