Ytterbium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-7030-10G 93-7030-2G
100209-452EA 607.2 USD
100209-452 100205-744
Ytterbium ≥99.9%, powder
Ytterbium
Formula: Yb
MW: 173.04 g/mol
Boiling Pt: 1427
Melting Pt: 824
Density: 6.97
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
UN: 3089
ADR: 4 III
Merck Index: 12,10239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR