Thulium(III) oxide ≥99.9% (Tm basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6904-1G 93-6904-5G
100209-200EA 89.17 USD
100209-200 100209-202
Thulium(III) oxide ≥99.9% (Tm basis)
Thulium (III) oxide
Formula: O₃Tm₂
MW: 385.87 g/mol
Melting Pt: 2410
Density: 8.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR