Terbium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS
Warning

93-6517-10G 93-6517-2G
100202-714EA 1057.29 USD
100202-714 100197-934
Terbium ≥99.9%, ingot
Terbium
Formula: Tb
MW: 158.93 g/mol
Boiling Pt: 3041 °C
Melting Pt: 1360 °C
Density: 8.234
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
Merck Index: 12,09300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR