Europium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS
Danger

93-6335-1G 93-6335-5G
100199-156EA 294.4 USD
100199-156 100205-898
Europium ≥99.9%, ingot
Europium
Formula: Eu
Melting Pt: 822 °C
Density: 5.24
MDL Number: MFCD00010992
CAS Number: 7440-53-1
EINECS: 231-161-7
UN: 3208
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR