Europium trinitrate hexahydrate ≥99.9% (Eu basis)

Supplier: 0000018203
Danger

93-6310-5G 93-6310-1G
100210-514EA 50.95 USD
100210-514 100203-546
Europium trinitrate hexahydrate ≥99.9% (Eu basis)
Europium trinitrate hexahydrate
Formula: EuN₃O₉·6H₂O
MW: 446.07 g/mol
Melting Pt: 85 °C
Density: 2.258 g/cm³ (20 °C)
MDL Number: MFCD00149700
CAS Number: 10031-53-5
EINECS: 233-380-3
UN: 1477
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR