Samarium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS
Warning

93-6240-50X50MM 93-6240-25X25MM
100196-992EA 1221.48 USD
100196-992 100199-154
Samarium ≥99.9%, foil
Samarium
Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900
Melting Pt: 1076.85
Density: 7.4
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR