Samarium ≥99.9%, ingot
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6237-100G
93-6237-25G
100202-466EA
743.08
USD
100202-466
100207-884
Samarium ≥99.9%, ingot
Samarium
Formula:
Sm MW: 150.36 g/mol Boiling Pt: 1900 Melting Pt: 1076.85 Density: 7.4 Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...