Samarium ≥99.9%, chips
Supplier: STREM CHEMICALS INC MS
93-6236-50G
93-6236-10G
100196-798EA
513.78
USD
100196-798
100196-796
Samarium ≥99.9%, chips
Samarium
Formula:
Sm MW: 150.36 g/mol Boiling Pt: 1900 Melting Pt: 1076.85 Density: 7.4 Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...