Cerium(IV) oxide ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Ceric oxide, Cerium dioxide

93-5816-50G 93-5816-1000G 93-5816-250G
100202-222EA 41.05 USD
100202-222 100202-220 100204-738
Cerium(IV) oxide ≥99.9% (Ce basis)
Cerium (IV) oxide
Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 7.132
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR