Barium oxide ≥99.5% (Ba basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-5651-100G 93-5651-25G
100196-988EA 849.23 USD
100196-988 100201-130
Barium oxide ≥99.5% (Ba basis)
Barium oxide
Formula: BaO
MW: 153.33 g/mol
Boiling Pt: ∼2000 °C (1013 hPa)
Melting Pt: 1923 °C
Density: 5.72
Storage Temperature: Ambient
MDL Number: MFCD00003453
CAS Number: 1304-28-5
EINECS: 215-127-9
UN: 1884
ADR: 6.1,III
Merck Index: 12,01015

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR