Cesium nitrate ≥99.99% (Cs basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Nitric acid cesium salt

93-5545-10G 93-5545-50G
100205-656EA 79.26 USD
100205-656 100203-518
Cesium nitrate ≥99.99% (Cs basis), PURATREM
Cesium nitrate
Formula: CsNO₃
MW: 194.91 g/mol
Boiling Pt: 737
Melting Pt: 414
Density: 3.685
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1 III
Merck Index: 12,02057

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR