Cesium bromide ≥99.9% (Cs basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cesiumbromide

93-5541-50G 93-5541-10G
100209-156EA 270.34 USD
100209-156 100197-898
Cesium bromide ≥99.9% (Cs basis)
Cesium bromide
Formula: BrCs
MW: 212.81
Boiling Pt: 1300
Melting Pt: 636
Density: 4.44
MDL Number: MFCD00010954
CAS Number: 7787-69-1
EINECS: 232-130-0
Merck Index: 13,02019

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR