Tellurium ≥99.8%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5222-500G
93-5222-100G
93-5222-25G
100197-892EA
706.28
USD
100197-892
100197-890
100207-104
Tellurium ≥99.8%, powder
Tellurium
Formula:
Te MW: 127.6 g/mol Boiling Pt: 990 Melting Pt: 450 Density: 6.24 |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3288 ADR: 6.1 III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...