Tin ≥99.8%, shot

Supplier: STREM CHEMICALS INC MS

93-5088-250G 93-5088-50G
100196-722EA 107.57 USD
100196-722 100202-188
Tin ≥99.8%, shot
Tin
Formula: Sn
Boiling Pt: 2260 °C
Melting Pt: 231.9 °C
Density: 7.31
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR