Tetrallyltin ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetrallylstannane, Tetraallyltin

93-5080-2G 93-5080-10G
100202-186EA 125.97 USD
100202-186 100202-806
Tetrallyltin ≥95%
Tetrallyltin
Formula: C₁₂H₂₀Sn
MW: 283
Density: 1.179
Flash Pt: 167 °F
MDL Number: MFCD00008637
CAS Number: 7393-43-3
EINECS: 230-987-5
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR