Tin (II) sulfate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tin sulfate, Tin(II) sulphate, Tin sulphate, Tin(II) sulfate, Stannous sulphate, Stannous sulfate
93-5044-500G
93-5044-100G
100196-712EA
107.57
USD
100196-712
100202-178
Tin (II) sulfate ≥95%
Tin (II) sulphate
Formula:
O₄SSn Melting Pt: > 360 °C Density: 4.15 g/cm³ (20 °C) |
MDL Number:
MFCD00011246 CAS Number: 7488-55-3 EINECS: 231-302-2 Merck Index: 12,08947 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...