Tin(IV) oxide ≥99.9% (Sn basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tin(iv)oxide, Stannic Oxide, Tin dioxide
93-5026-500G
93-5026-100G
100205-620EA
181.17
USD
100205-620
100208-572
Tin(IV) oxide ≥99.9% (Sn basis)
Tin (IV) oxide
Formula:
O₂Sn MW: 150.71 g/mol Boiling Pt: ∼2500 °C (1013 hPa) Melting Pt: 1624.85 Density: 7.0 |
MDL Number:
MFCD00011244 CAS Number: 18282-10-5 EINECS: 242-159-0 Merck Index: 14,08778 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...