Indium ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-4941-5G 93-4941-25G
100208-726EA 237.78 USD
100208-726 100205-616
Indium ≥99.999%, powder
Indium
Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.31
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4 III
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR