Cadmium sulfide ≥99.999% (Cd basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cadmium (II) sulphide
, Cadmium (II) sulfide
93-4840-50G
93-4840-10G
100205-608EA
277.42
USD
100205-608
100196-696
Cadmium sulfide ≥99.999% (Cd basis), PURATREM
Cadmium sulphide
Formula:
CdS MW: 144.48 g/mol Melting Pt: 1750 °C Density: 4.82 |
MDL Number:
MFCD00010922 CAS Number: 1306-23-6 EINECS: 215-147-8 UN: 2570 ADR: 6.1,III Merck Index: 12,01666 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...