Cadmium sulfide ≥99.9% (Cd basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cadmium (II) sulphideCadmium (II) sulfide

93-4822-100G 93-4822-25G 93-4822-500G
100196-692EA 130.22 USD
100196-692 100199-112 100205-602
Cadmium sulfide ≥99.9% (Cd basis)
Cadmium sulphide
Formula: CdS
MW: 144.48 g/mol
Melting Pt: 1750 °C
Density: 4.82
MDL Number: MFCD00010922
CAS Number: 1306-23-6
EINECS: 215-147-8
UN: 2570
ADR: 6.1,III
Merck Index: 12,01666

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR